missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Nephronophthisis Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30792-50ug
This item is not returnable.
View return policy
Description
Nephronophthisis Polyclonal antibody specifically detects Nephronophthisis in Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| Nephronophthisis | |
| Polyclonal | |
| Western Blot : 2-10 μg/mL, Immunoprecipitation : 10-20 μg/mL | |
| FLJ97602, JBTS4NPH1Juvenile nephronophthisis 1 protein, nephrocystin 1, nephrocystin-1, nephronophthisis 1 (juvenile), SLSN1 | |
| Recombinant protein corresponding to the amino acid 612-691 of the C terminus region and having a molecular weight of 90 kD | |
| 50 μg | |
| Cell Biology, Signal Transduction | |
| 4867 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur