missing translation for 'onlineSavingsMsg'
Learn More

Neomycin phosphotransferase II Antibody (4B4D1), Novus Biologicals™

Mouse Monoclonal Antibody

£134.00

Specifications

Antigen Neomycin phosphotransferase II
Clone 4B4D1
Dilution Western Blot 1:500-1:2000, ELISA 1:10000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18581173
View Documents
Novus Biologicals
NB110-60487-0.025ml
0.025 mL
£134.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Neomycin phosphotransferase II Monoclonal specifically detects Neomycin phosphotransferase II in Non-species specific, Bacteria samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Neomycin phosphotransferase II
Western Blot 1:500-1:2000, ELISA 1:10000
Monoclonal
Ascites
RUO
NPT2, NPTII
IgG2a
Unpurified
Transfected or transformed cells/organisms (plants, animals) carrying NPTII gene encoding neomycin phosphotransferase II enzyme. Escherichia coli (E. coli).
4B4D1
Western Blot
Unconjugated
Mouse
Non-species specific
Ni-NTA purified truncated recombinant NPT-II expressed in E. Coli strain BL21 (DE3)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.