missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neomycin phosphotransferase II Antibody (4B4D1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-60487-0.025ml
This item is not returnable.
View return policy
Description
Neomycin phosphotransferase II Monoclonal specifically detects Neomycin phosphotransferase II in Non-species specific, Bacteria samples. It is validated for Western Blot, ELISA.
Specifications
| Neomycin phosphotransferase II | |
| Monoclonal | |
| Unconjugated | |
| NPT2, NPTII | |
| Ni-NTA purified truncated recombinant NPT-II expressed in E. Coli strain BL21 (DE3) | |
| 0.025 mL | |
| Primary | |
| Non-species specific | |
| Ascites |
| Western Blot | |
| 4B4D1 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Transfected or transformed cells/organisms (plants, animals) carrying NPTII gene encoding neomycin phosphotransferase II enzyme. Escherichia coli (E. coli). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction