missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NELF-E Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £513.00
Specifications
| Antigen | NELF-E |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18266542
|
Novus Biologicals
NBP2-55487 |
100 μL |
£513.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18693538
|
Novus Biologicals
NBP2-55487-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NELF-E Polyclonal specifically detects NELF-E in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| NELF-E | |
| Polyclonal | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| D6S45, negative elongation factor E, negative elongation factor polypeptide E, NELFE, NELF-ERDmajor histocompatibility complex gene RD, nuclear protein, RD RNA binding protein, RD RNA-binding protein, RDP, RNA-binding protein RD | |
| RDBP | |
| IgG | |
| Affinity Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 7936 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:VYGEDMTPTLLRGAFSPFGNIIDLSMDPPRNCAFVTYEKMESADQAVAELNGTQVESVQLKVNIARKQPMLDAATGKSVWGSLAVQNSPKGCHRDKRTQIVYSDDVYKE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title