missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NELF-E Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-57127
This item is not returnable.
View return policy
Description
NELF-E Polyclonal specifically detects NELF-E in Human samples. It is validated for Western Blot.
Specifications
| NELF-E | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| D6S45, negative elongation factor E, negative elongation factor polypeptide E, NELFE, NELF-ERDmajor histocompatibility complex gene RD, nuclear protein, RD RNA binding protein, RD RNA-binding protein, RDP, RNA-binding protein RD | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Mouse: 100%; Canine: 92%; Guinea pig: 92%; Pig: 92%; Rabbit: 92%; Rat: 92%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| P18615 | |
| RDBP | |
| Synthetic peptides corresponding to RDBP(RD RNA binding protein) The peptide sequence was selected from the N terminal of RDBP. Peptide sequence QSSSSTTSQGGVKRSLSEQPVMDTATATEQAKQLVKSGAISAIKAETKNS. | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 7936 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction