missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NEK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £488.00
Specifications
| Antigen | NEK2 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18238344
|
Novus Biologicals
NBP2-57424 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18667938
|
Novus Biologicals
NBP2-57424-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NEK2 Polyclonal specifically detects NEK2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| NEK2 | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication, Core ESC Like Genes, Mitotic Regulators, Stem Cell Markers | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 4751 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:FSQKELAGKIREGKFRRIPYRYSDELNEIITRMLNLKDYHRPSVEEILENPLIADLVADEQRRNLERRGRQLGEPE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| EC 2.7.11, EC 2.7.11.1, HSPK 21, NEK2AHsPK21, Never in mitosis A-related kinase 2, NIMA (never in mitosis gene a)-related kinase 2, NimA-like protein kinase 1, NimA-related protein kinase 2, NLK1serine/threonine-protein kinase Nek2 | |
| NEK2 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title