missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEK11 (FLJ23495) Recombinant Human Protein
NEK11 (FLJ23495) belongs to the NIMA family of kinases.
£434.00 - £6900.00
Specifications
Accession Number | NP_079076 |
---|---|
Formulation | 50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol |
Molecular Weight (g/mol) | 101.6kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Regulatory Status | RUO - research use only |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10547523
|
Invitrogen™
PV4647 |
10μg |
£434.00
10µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15418663
|
Invitrogen™
PR8182A |
100μg |
£1602.00
100µg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15418743
|
Invitrogen™
PV4648 |
1mg |
£6900.00
1mg |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
NEK11 (FLJ23495) is involved in DNA replication and genotoxic stress response.
Specifications
NP_079076 | |
101.6kDa | |
RUO - research use only | |
NEK Kinase Family | |
Recombinant | |
Baculovirus |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
NEK11 (FLJ23495) | |
Human | |
GST Tag |