missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEK11 (FLJ23495) Recombinant Human Protein
NEK11 (FLJ23495) belongs to the NIMA family of kinases.
Brand: Invitrogen™ PV4647
Code : 50
Additional Details : Weight : 0.50000kg
Description
NEK11 (FLJ23495) is involved in DNA replication and genotoxic stress response.
Specifications
NP_079076 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
101.6kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length, Recombinant | |
NIMA (never in mitosis gene a)- related kinase 11 | |
Human | |
GST Tag | |
Kinases & Inhibitors |
Cell Analysis, DNA Replication and Repair, Nuclear Receptor Assay | |
79858 | |
10μg | |
RUO - research use only | |
NEK11 (FLJ23495) | |
NEK Kinase Family | |
Recombinant | |
Baculovirus | |
Serine/Threonine Kinases |