missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human NEK1, GST Tag Recombinant Protein

NEK1 is a serine/threonine protein kinase, related to the NIMA cell cycle regulator, and is highly expressed in meiotic germ cells.

Brand:  Invitrogen™ PR7201A

Code : NEW

Additional Details : Weight : 0.00900kg

 View more versions of this product

Product Code. 12644770

  • £2555.00 / 100µg
In Stock
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The NEK1 kinase may function early in the DNA damage response pathway. It may also participate in polycystic kidney disease and neural cell development.

Specifications

Specifications

NP_036356
85.3 kDa
NEK1
Human
GST Tag
Ser⁄Thr Kinases: Other
Baculovirus
Frozen
Kinase Assay
100μg
NEK Kinase Family
Recombinant
Storage Buffer: 50 mM Tris pH 7.5, 150 mM NaCl, 0.5 mM EDTA, 0.02% Triton X-100, 2 mM DTT, 50% Glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Dry Ice
Catalytic
Drug Development
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers