missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human NEK1, GST Tag Recombinant Protein
NEK1 is a serine/threonine protein kinase, related to the NIMA cell cycle regulator, and is highly expressed in meiotic germ cells.
£2555.00 - £12060.00
Specifications
Accession Number | NP_036356 |
---|---|
Gene Alias | NEK1 |
KinaseFamily | NEK Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12644770
|
Invitrogen™
PR7201A |
100μg |
£2555.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10332303
|
Invitrogen™
PV4282 |
1mg |
£12060.00
1mg |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The NEK1 kinase may function early in the DNA damage response pathway. It may also participate in polycystic kidney disease and neural cell development.
Specifications
NP_036356 | |
NEK Kinase Family | |
Recombinant | |
Baculovirus |
NEK1 | |
Human | |
GST Tag | |
Frozen |