Learn More
Invitrogen™ Human NEK1, GST Tag Recombinant Protein
NEK1 is a serine/threonine protein kinase, related to the NIMA cell cycle regulator, and is highly expressed in meiotic germ cells.
Brand: Invitrogen™ PV4282
Code : NEW
Additional Details : Weight : 0.00900kg
Description
The NEK1 kinase may function early in the DNA damage response pathway. It may also participate in polycystic kidney disease and neural cell development.
Specifications
NP_036356 | |
50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
1mg | |
RUO - research use only | |
NEK1 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
85.3kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
NEK Kinase Family | |
Recombinant | |
Baculovirus |