missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nectin-3/PVRL3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£183.00 - £437.00
Specifications
| Antigen | Nectin-3/PVRL3 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.5 - 4 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18263511
|
Novus Biologicals
NBP1-45736-0.025MG |
0.025 mg |
£183.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18229224
|
Novus Biologicals
NBP1-45736 |
0.1 mg |
£437.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nectin-3/PVRL3 Polyclonal specifically detects Nectin-3/PVRL3 in Mouse, Rat samples. It is validated for Western Blot.Specifications
| Nectin-3/PVRL3 | |
| Western Blot 0.5 - 4 ug/mL | |
| Polyclonal | |
| Rabbit | |
| Rat, Mouse | |
| CD113 antigen, CDw113FLJ90624, DKFZp566B0846, nectin 3, nectin-3, poliovirus receptor-related 3, poliovirus receptor-related protein 3, PPR3, PRR3CD113, PVRR3 | |
| NECTIN3 | |
| IgG | |
| Affinity Purified |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9NQS3 | |
| 25945 | |
| Synthetic peptide corresponding to residues surrounding amino acids 536 of mouse Nectin-3. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title