missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nectin-3/PVRL3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-45736
This item is not returnable.
View return policy
Description
Nectin-3/PVRL3 Polyclonal specifically detects Nectin-3/PVRL3 in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Nectin-3/PVRL3 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2 ), 5 mM EDTA, 30% glycerol, 0.5% BSA with 0.03% Proclin 300 | |
| CD113 antigen, CDw113FLJ90624, DKFZp566B0846, nectin 3, nectin-3, poliovirus receptor-related 3, poliovirus receptor-related protein 3, PPR3, PRR3CD113, PVRR3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 4 ug/mL | |
| Q9NQS3 | |
| NECTIN3 | |
| Synthetic peptide corresponding to residues surrounding amino acids 536 of mouse Nectin-3. | |
| 0.1 mg | |
| Immunology | |
| 25945 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction