missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NECAP2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£366.00
Specifications
| Antigen | NECAP2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18637091
|
Novus Biologicals
NBP2-93292-0.02ml |
0.02 mL |
N/A
|
N/A | |||||
|
18657050
|
Novus Biologicals
NBP2-93292-0.1ml |
0.1 mL |
£366.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NECAP2 Polyclonal antibody specifically detects NECAP2 in Mouse samples. It is validated for Western BlotSpecifications
| NECAP2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 55707 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| adaptin ear-binding coat-associated protein 2, adaptin-ear-binding coat-associated protein 2, FLJ10420, NECAP endocytosis associated 2, NECAP endocytosis-associated protein 2, NECAP-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 194-263 of human NECAP2 (NP_060560.1). GKTSTLIPPPGEQLAVGGSLVQPAVAPSSGGAPVPWPQPNPATADIWGDFTKSTGSTSSQTQPGTGWVQF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title