missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NECA
GREENER_CHOICE

Product Code. 30001278
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001278

Brand: Tocris Bioscience™ 1691/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity adenosine receptor agonist

High affinity adenosine receptor agonist (Ki values are 6.2, 14, and 20 nM for human A3, A1 and A2A receptors respectively; EC50 = 2.4 μM for human A2B). Inhibits platelet aggregation and is centrally active in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35920-39-9
Molecular Formula C12H16N6O4

Specifications

Quantity 10 mg
Formula Weight 308.3
Purity 98%
Molecular Weight (g/mol) 308.30
Chemical Name or Material NECA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.