missing translation for 'onlineSavingsMsg'
Learn More

NDUFS8 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£381.00

Specifications

Antigen NDUFS8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18270106
View Documents
Novus Biologicals
NBP1-79907
100 μL
£381.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NDUFS8 Polyclonal specifically detects NDUFS8 in Human samples. It is validated for Western Blot.
Specifications

Specifications

NDUFS8
Polyclonal
Rabbit
NP_002487
4728
The immunogen for this antibody is NDUFS8. Peptide sequence: CKLCEAICPAQAITIEAEPRADGSRRTTRYDIDMTKCIYCGFCQEACPVD
Primary
Western Blot
Unconjugated
RUO
CI-23k, CI-23kD, CI23KD, complex I 23kDa subunit, Complex I-23kD, EC 1.6.5.3, EC 1.6.99.3, EC 1.6.99.5, NADH dehydrogenase (ubiquinone) Fe-S protein 8 (23kD) (NADH-coenzyme Qreductase), NADH dehydrogenase (ubiquinone) Fe-S protein 8, 23kDa (NADH-coenzyme Qreductase), NADH dehydrogenase [ubiquinone] iron-sulfur protein 8, mitochondrial, NADH-ubiquinone oxidoreductase 23 kDa subunit, TYKY, TYKY subunit
NDUFS8
IgG
23 kDa
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.