missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDUFB4 Recombinant Rabbit Monoclonal Antibody (24GB500)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£126.00 - £434.00

Specifications

Antigen NDUFB4
Clone 24GB500
Content And Storage -20°C
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30275904
GREENER_CHOICE
View Documents
Invitrogen™
MA557103
100 μL
£434.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30276611
View Documents
Invitrogen™
MA557104
20 μL
£126.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NDUFB4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

The protein encoded by this gene belongs to the complex I NDUFB4 subunit family. Mammalian complex I is composed of 45 different subunits. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications

Specifications

NDUFB4
-20°C
Recombinant Monoclonal
Liquid
IgG
Human, Mouse, Rat
NDUFB4
0610006N12Rik; 1300010H20Rik; B15; B15 subunit of the NADH: ubiquinone oxidoreductase complex; CI-B15; complex I B15 subunit; complex I-B15; MGC5105; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 4; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 4, 15kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 4; NADH:ubiquinone oxidoreductase B15 subunit; NADH:ubiquinone oxidoreductase subunit B4; NADH-ubiquinone oxidoreductase B15 subunit; Ndufb4; similar to B15 subunit of the NADH: ubiquinone oxidoreductase complex
NDUFB4
Primary
Affinity chromatography
24GB500
Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O95168, Q9CQC7
288088, 4710, 68194
A synthesized peptide derived from human NDUFB4 (aa 1-129).
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.