missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFB4 Recombinant Rabbit Monoclonal Antibody (24GB500)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557103
This item is not returnable.
View return policy
Description
NDUFB4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The protein encoded by this gene belongs to the complex I NDUFB4 subunit family. Mammalian complex I is composed of 45 different subunits. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
| NDUFB4 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| NDUFB4 | |
| 0610006N12Rik; 1300010H20Rik; B15; B15 subunit of the NADH: ubiquinone oxidoreductase complex; CI-B15; complex I B15 subunit; complex I-B15; MGC5105; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 4; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 4, 15kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 4; NADH:ubiquinone oxidoreductase B15 subunit; NADH:ubiquinone oxidoreductase subunit B4; NADH-ubiquinone oxidoreductase B15 subunit; Ndufb4; similar to B15 subunit of the NADH: ubiquinone oxidoreductase complex | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 288088, 4710, 68194 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 24GB500 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O95168, Q9CQC7 | |
| NDUFB4 | |
| A synthesized peptide derived from human NDUFB4 (aa 1-129). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur