missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA9 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54761
This item is not returnable.
View return policy
Description
NDUFA9 Polyclonal specifically detects NDUFA9 in Human samples. It is validated for Western Blot.
Specifications
| NDUFA9 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| CC6, CI39k, CI-39k, CI-39kD, complex I 39kDa subunit, Complex I-39kD, MGC111043, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9 (39kD), NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9, 39kDa, NADH dehydrogenase (ubiquinone) Fe-S protein 2-like (NADH-coenzyme Q reductase), NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 9, mitochondrial, NADH-ubiquinone oxidoreductase 39 kDa subunit, NDUFS2L, SDR22E1, short chain dehydrogenase/reductase family 22E, member 1 | |
| Rabbit | |
| 42 kDa | |
| 100 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 4704 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q16795 | |
| NDUFA9 | |
| Synthetic peptides corresponding to NDUFA9(NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 9, 39kDa) The peptide sequence was selected from the N terminal of NDUFA9. Peptide sequence QLHHALMPHGKGGRSSVSGIVATVFGATGFLGRYVVNHLGRMGSQVIIPY. | |
| Affinity purified | |
| RUO | |
| Primary | |
| This product is specific to Subunit or Isoform: 9, mitochondrial. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction