missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFA13 Recombinant Rabbit Monoclonal Antibody (24GB600)

Rabbit Recombinant Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | NDUFA13 |
|---|---|
| Clone | 24GB600 |
| Concentration | 1.7 mg/mL |
| Content And Storage | -20°C |
| Applications | Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275353
![]() |
Invitrogen™
MA557496 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275838
|
Invitrogen™
MA557497 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NDUFA13 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
A novel gene associated with Retinoid-IFN-induced Mortality (GRIM) GRIM-19 gene was identified. Antisense expression of GRIM-19 confers a strong resistance against IFN/RA-induced death by reducing the intracellular levels of GRIM-19 protein. Overexpression of GRIM-19 enhances cell death in response to IFN/RA. GRIM-19 is primarily a nuclear protein whose expression is induced by the IFN/RA combination. These data indicate that GRIM-19 is a novel cell death-regulatory molecule.Specifications
| NDUFA13 | |
| 1.7 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9P0J0 | |
| 51079 | |
| A synthesized peptide derived from human GRIM19 (aa 60-100). | |
| Antibody |
| 24GB600 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Ndufa13 | |
| 2700054G14Rik; AU022060; B16.6; CDA016; Cell d; Cell death regulatory protein GRIM-19; cell death-regulatory protein GRIM19; CGI-39; CI-B16.6; complex I B16.6 subunit; complex I-B16.6; FLJ58045; FLJ59191; gene associated with retinoic and IFN-induced mortality 19 protein; gene associated with retinoic and interferon-induced mortality 19 protein; Gene associated with retinoic-interferon-induced mortality 19 protein; genes associated with retinoid-IFN-induced mortality 19; GRIM19; GRIM-19; LOC100911483; mitochondrial NADH:ubiquinone oxidoreductase B16.6 subunit; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 13-like; NADH:ubiquinone oxidoreductase subunit A13; NADH-ubiquinone oxidoreductase B16.6 subunit; Ndufa13; YjeF N-terminal domain-containing protein 3; YjeF_N3; Yjefn3 | |
| Ndufa13 | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title