Learn More
NDP52 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | NDP52 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence, Flow Cytometry (Intracellular Staining) |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18034815
|
Proteintech
CL488-12229-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18099375
|
Proteintech
CL488-12229-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NDP52 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NDP52. This antibody recognizes human, mouse, rat antigen. The NDP52 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
| NDP52 | |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| Liquid | |
| Human, Mouse, Rat | |
| CALCOCO2, NDP52, Nuclear domain 10 protein 52, Nuclear dot protein 52 | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 10241 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.