Learn More
NDP52 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-12229-100UL
 View more versions of this product
Product Code. 18034815
  • £375.00 / 100µL
Description
The NDP52 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NDP52. This antibody recognizes human, mouse, rat antigen. The NDP52 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
NDP52
1000 μg/mL
CALCOCO2, NDP52, Nuclear domain 10 protein 52, Nuclear dot protein 52
100 μL
10241
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence, Flow Cytometry (Intracellular Staining)
CoraLite Plus 488
Rabbit
Primary
Human, Mouse, Rat
150 kDa