missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ND6 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£186.00 - £386.00
Specifications
| Antigen | ND6 |
|---|---|
| Dilution | Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18690812
|
Novus Biologicals
NBP2-94464-0.02ml |
0.02 mL |
£186.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18634692
|
Novus Biologicals
NBP2-94464-0.1ml |
0.1 mL |
£386.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ND6 Polyclonal antibody specifically detects ND6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| ND6 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 4541 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| mitochondrially encoded NADH dehydrogenase 6, MTND6, MT-ND6, NADH dehydrogenase subunit 6, NADH dehydrogenase, subunit 6 (complex I), NADH6 | |
| A synthetic peptide corresponding to a sequence within amino acids 80-174 of human ND6 (YP_003024037.1). LIREDPIGAGALYDYGR | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title