missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ND6 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94464-0.02ml
This item is not returnable.
View return policy
Description
ND6 Polyclonal antibody specifically detects ND6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ND6 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
| mitochondrially encoded NADH dehydrogenase 6, MTND6, MT-ND6, NADH dehydrogenase subunit 6, NADH dehydrogenase, subunit 6 (complex I), NADH6 | |
| A synthetic peptide corresponding to a sequence within amino acids 80-174 of human ND6 (YP_003024037.1). LIREDPIGAGALYDYGR | |
| 0.02 mL | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 4541 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction