missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCS1 Mouse anti-Human, Unconjugated, Clone: 2F10C8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | NCS1 |
|---|---|
| Clone | 2F10C8 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16820346
|
Proteintech
67616-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16810346
|
Proteintech
67616-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NCS1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NCS1. This antibody recognizes human, mouse, rat, pig antigen. The NCS1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| NCS1 | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 23413 | |
| Primary |
| 2F10C8 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Human, Mouse, Rat, Pig | |
| IgG2a |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title