Learn More
NCS1 Mouse anti-Human, Unconjugated, Clone: 2F10C8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67616-1-IG-20UL
 View more versions of this product
Product Code. 16820346
  • £135.00 / 20µL
Description
The NCS1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NCS1. This antibody recognizes human, mouse, rat, pig antigen. The NCS1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
NCS1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence
2F10C8
Unconjugated
20 μL
23413
IgG2a