missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOA7 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33462-20ul
This item is not returnable.
View return policy
Description
NCOA7 Monoclonal antibody specifically detects NCOA7 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| NCOA7 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| 140 kDa estrogen receptor-associated protein, dJ187J11.3, ERAP140Nbla00052, ESNA1, Estrogen nuclear receptor coactivator 1, estrogen receptor associated protein 140 kDa, FLJ45605, MGC88425, Nbla10993, nuclear receptor coactivator 7, putative protein product of Nbla00052, putative protein product of Nbla10993 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 777-942 of human NCOA7.3).,, Sequence:, LRPHSALLENMHIEQLARRLPARVQGYPWRLAYSTLEHGTSLKTLYRKSASLDSPVLLVIKDMDNQIFGAYATHPFKFSDHYYGTGETFLYTFSPHFKVFKWSGENSYFINGDISSLELGGGGGRFGLWLDADLYHGRSNSCSTFNNDILSKKEDFIVQDLEVWAFD | |
| 20 μL | |
| Signal Transduction | |
| 135112 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction