missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCOA7 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
£161.00 - £383.00
Specifications
| Antigen | NCOA7 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229805
|
Novus Biologicals
NBP3-33462-100ul |
100 μL |
£383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228656
|
Novus Biologicals
NBP3-33462-20ul |
20 μL |
£161.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NCOA7 Monoclonal antibody specifically detects NCOA7 in Human,Mouse samples. It is validated for ELISA,Western BlotSpecifications
| NCOA7 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| 135112 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 140 kDa estrogen receptor-associated protein, dJ187J11.3, ERAP140Nbla00052, ESNA1, Estrogen nuclear receptor coactivator 1, estrogen receptor associated protein 140 kDa, FLJ45605, MGC88425, Nbla10993, nuclear receptor coactivator 7, putative protein product of Nbla00052, putative protein product of Nbla10993 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 777-942 of human NCOA7.3).,, Sequence:, LRPHSALLENMHIEQLARRLPARVQGYPWRLAYSTLEHGTSLKTLYRKSASLDSPVLLVIKDMDNQIFGAYATHPFKFSDHYYGTGETFLYTFSPHFKVFKWSGENSYFINGDISSLELGGGGGRFGLWLDADLYHGRSNSCSTFNNDILSKKEDFIVQDLEVWAFD | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title