missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£435.00
Specifications
| Antigen | NCK1 |
|---|---|
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
NCK1 Polyclonal specifically detects NCK1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| NCK1 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Cell Cycle and Replication | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 4690 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MAEEVVVVAKFDYVAQQEQELDIKKNERLWLLDDSKSWWRVRNSMNKTGFVPSNYVERKNSARKASIVKNLKDTLGIGKVKRKPSVPDSASPADDSFVDPGERLYDLNMPAYVKFNYMAEREDELSLIKGTKVIVMEKCSDGWWRGSYNG | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| cytoplasmic protein NCK1, MGC12668, NCK adaptor protein 1melanoma NCK protein, nck-1, NCKalpha, NCKNCK tyrosine kinase, non-catalytic region of tyrosine kinase, SH2/SH3 adaptor protein NCK-alpha | |
| NCK1 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title