missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-55255
This item is not returnable.
View return policy
Description
NCK1 Polyclonal specifically detects NCK1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| NCK1 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunocytochemistry/Immunofluorescence 1-4 ug/ml | |
| cytoplasmic protein NCK1, MGC12668, NCK adaptor protein 1melanoma NCK protein, nck-1, NCKalpha, NCKNCK tyrosine kinase, non-catalytic region of tyrosine kinase, SH2/SH3 adaptor protein NCK-alpha | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| NCK1 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:MAEEVVVVAKFDYVAQQEQELDIKKNERLWLLDDSKSWWRVRNSMNKTGFVPSNYVERKNSARKASIVKNLKDTLGIGKVKRKPSVPDSASPADDSFVDPGERLYDLNMPAYVKFNYMAEREDELSLIKGTKVIVMEKCSDGWWRGSYNG | |
| 100 μL | |
| Cancer, Cell Cycle and Replication | |
| 4690 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction