missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Natural Killer Cell Antibody (H25) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £302.00
Specifications
| Antigen | Natural Killer Cell |
|---|---|
| Clone | H25 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 |
| Applications | Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18618846
|
Novus Biologicals
NBP2-50526-0.025ml |
0.025 mL |
£150.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18658846
|
Novus Biologicals
NBP2-50526 |
0.1 mL |
£302.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Natural Killer Cell Monoclonal antibody specifically detects Natural Killer Cell in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Radioimmune AssaysSpecifications
| Natural Killer Cell | |
| 1 mg/mL | |
| Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA) | |
| Unconjugated | |
| Mouse | |
| Immunology | |
| PBS | |
| HSE-2 cell line | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| H25 | |
| Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| NK Cell | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title