missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Natural Killer Cell Antibody (H25) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50526-0.025ml
This item is not returnable.
View return policy
Description
Natural Killer Cell Monoclonal antibody specifically detects Natural Killer Cell in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Radioimmune Assays
Specifications
| Natural Killer Cell | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 | |
| NK Cell | |
| HSE-2 cell line | |
| 0.025 mL | |
| Immunology | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Radioimmune Assays (RIA) | |
| H25 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction