Learn More
Thermo Scientific™ National™ Target™ Kim-Snap™ Closures
Preassembled with septa for use with Target Snap/Crimp LoVial™ Vials
Brand: Thermo Scientific™ C4010-1296VO
Code : NEW
Additional Details : Weight : 1.00000kg
Description
- Effective evaporation resistance—apply with simple thumb pressure
- Replace aluminum crimp seals and end tedious crimping/de-crimping
Specifications
PTFE/Silicone | |
Black | |
Target Snap/Crimp LoVial Vials |