missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ National™ Target™ Kim-Snap™ Closures

Product Code. p-2363687 Shop All Thermo Scientific Products
Click to view available options
Color:
Black
Yellow
Material (Liner):
PTFE/Rubber
PTFE/Silicone
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 13120963

Brand: Thermo Scientific™ C401151Y

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Preassembled with septa for use with Target Snap/Crimp LoVial™ Vials

  • Effective evaporation resistance—apply with simple thumb pressure
  • Replace aluminum crimp seals and end tedious crimping/de-crimping

TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) PTFE/Rubber
Color Yellow
Material Polypropylene
Closure Size 11 mm
For Use With (Equipment) Target Snap/Crimp LoVial Vials
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.