Learn More
NAT10 Rabbit, anti-Human, CoraLite Plus 488, Clone: 5B9, Proteintech
Rabbit Recombinant Monoclonal Antibody
£135.00 - £375.00
Specifications
| Antigen | NAT10 |
|---|---|
| Clone | 5B9 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18007815
|
Proteintech
CL488-82585-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18099465
|
Proteintech
CL488-82585-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The NAT10 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human NAT10. This antibody recognizes human antigen. The NAT10 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| NAT10 | |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 55226 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
| 5B9 | |
| Immunofluorescence | |
| Liquid | |
| Human | |
| ALP, DKFZp434C116, FLJ10774, FLJ12179, FLJ23850, KIAA1709, N acetyltransferase 10, NAT10 | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.