Learn More
NAT10 Rabbit, anti-Human, CoraLite Plus 488, Clone: 5B9, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech CL488-82585-20UL
Description
The NAT10 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human NAT10. This antibody recognizes human antigen. The NAT10 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| NAT10 | |
| 5B9 | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Immunofluorescence | |
| 1000 μg/mL | |
| ALP, DKFZp434C116, FLJ10774, FLJ12179, FLJ23850, KIAA1709, N acetyltransferase 10, NAT10 | |
| 20 μL | |
| 55226 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |