missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Microbiology Bottle, Narrow Neck, Unground

Product Code. 11619858 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
10,000 mL
100 mL
1000 mL
20,000 mL
2000 mL
25 mL
250 mL
50 mL
500 mL
5000 mL
Unit Size:
Each
Pack of 10
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11619858 1000 mL Pack of 10
10246792 5000 mL Each
10655331 10,000 mL Each
11609858 500 mL Pack of 10
12810804 20,000 mL Each
11699848 100 mL Pack of 10
11629858 2000 mL Pack of 10
11777961 250 mL Pack of 10
11679848 25 mL Pack of 10
11689848 50 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 11619858 Supplier DWK Life Sciences Supplier No. 211615409
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Culture bottles with a narrow neck for use with rubber stoppers

The culture bottles are moulded from DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the preparation of media or culturing of microorganisms. The bottles comply with ISO 4796-2 and feature a narrow conical unground neck for use with rubber stoppers. Suitable for autoclaving at 121 or 134°C or dry heat sterilization.

  • Narrow neck for rubber stoppers
  • Retrace code
  • Autoclavable at 121°C or 134°C
  • Supplied without stopper
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
For Use With (Application) Autoclaving of media and reagents; Preparation of media and reagents; Culturing vessel
Includes With print
Material Borosilicate Glass (3.3)
Quantity 10/Pk.
Diameter (Metric) 107 mm
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.