missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences DURAN™ Microbiology Bottle, Narrow Neck, Unground
Shop All DWK Life Sciences ProductsDescription
The culture bottles are moulded from DURAN™ borosilicate 3.3 Type 1 glass, and are suitable for the preparation of media or culturing of microorganisms. The bottles comply with ISO 4796-2 and feature a narrow conical unground neck for use with rubber stoppers. Suitable for autoclaving at 121 or 134°C or dry heat sterilization.
- Narrow neck for rubber stoppers
- Retrace code
- Autoclavable at 121°C or 134°C
- Supplied without stopper
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| For Use With (Application) | Autoclaving of media and reagents; Preparation of media and reagents; Culturing vessel |
| Includes | With print |
| Material | Borosilicate Glass (3.3) |
| Quantity | 10/Pk. |
| Diameter (Metric) | 86 mm |
| Autoclavable | Autoclavable |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?