missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences DURAN™ Round Bottom Flask, narrow neck, with beaded rim, non-DIN ISO size

Product Code. 10252141 Shop All DWK Life Sciences Products
Change view
Click to view available options
Capacity (Metric):
12,000 mL
20,000 mL
2000 mL
3000 mL
5000 mL
6000 mL
Unit Size:
Each
Pack of 10
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10252141 6000 mL Each
11796073 2000 mL Pack of 10
10797561 3000 mL Each
10622581 5000 mL Each
10579641 20,000 mL Each
10736721 12,000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10252141 Supplier DWK Life Sciences Supplier No. 217217709
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Due to their spherical shape, round bottom flasks are used for heating liquids, among other things.

Abdominal vessels are called flasks in the laboratory and, in addition to beakers, are other universally applicable vessels. They are used e.g. for distillation or for heating liquids. Flasks are available in various designs: narrow and wide neck, with or without ground joint, as round or flat bottom flask. Depending on the application, the required flask with its specific components is selected.

  • The geometry permits very uniform heating
  • Fired-on, highly durable white ceramic
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Base 236 mm
Capacity (Metric) 6000 mL
Material Borosilicate Glass (3.3)
For Use With (Application) Suitable for heating mantles; Heating; Mixing several components; Sizes 2000, 3000, 5000, 6000, 12000 and 20000 mL non-DIN ISO size, size 12000 mL non-DIN ISO size, conforms ASTM E 1403 and Size 20000 mL conforms ASTM E 1403
No. per Pack 1/Pk.
Product Type Non-DIN/ISO size
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.