missing translation for 'onlineSavingsMsg'
Learn More

Naringin hydrate, 98%, Thermo Scientific Chemicals

Catalog Number 11338275
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
132203-74-8
C27H32O14
580.54
MFCD00148888,MFCD00149445,MFCD01461988
DFPMSGMNTNDNHN-ZPHOTFPESA-N
74787988
7-[(2S,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2OC2=CC(O)=C3C(=O)C[C@H](OC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@H](O)[C@H]1O
This item is not returnable. View return policy
132203-74-8
C27H32O14
580.54
MFCD00148888,MFCD00149445,MFCD01461988
DFPMSGMNTNDNHN-ZPHOTFPESA-N
74787988
7-[(2S,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-[(2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-2-yl]oxy-5-hydroxy-2-(4-hydroxyphenyl)-2,3-dihydrochromen-4-one
C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2OC2=CC(O)=C3C(=O)C[C@H](OC3=C2)C2=CC=C(O)C=C2)[C@H](O)[C@H](O)[C@H]1O

A citrus bioflavanoid that inhibits cytochrome P450 monooxygenase activity

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∽166°C (decomposition)
Quantity 50 g
Beilstein 102012
Merck Index 14,6424
Optical Rotation −90° (c=1 in Ethanol)
Formula Weight 580.55 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Naringin hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'