missing translation for 'onlineSavingsMsg'
Learn More

Naringenin 7-O-beta-D-Glucuronide Sodium Salt (Mixture of Diastereomers), TRC

Product Code. 30138362
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138362 10 mg 10mg
30011811 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30138362 Supplier Toronto Research Chemicals Supplier No. TRCN37896510MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C21H19NaO11
Molecular Weight (g/mol) 470.36
Synonym (2S)-3,4-Dihydro-5-hydroxy-2-(4-hydroxyphenyl)-4-oxo-2H-1-benzopyran-7-yl β-D-Glucopyranosiduronic Acid Sodium Salt
IUPAC Name sodium (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-((5-hydroxy-2-(4-hydroxyphenyl)-4-oxochroman-7-yl)oxy)tetrahydro-2H-pyran-2-carboxylate
SMILES OC1=CC(O[C@H]2[C@H](O)[C@@H](O)[C@H](O)[C@@H](C(O[Na])=O)O2)=CC3=C1C(CC(C4=CC=C(O)C=C4)O3)=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H20O11.Na/c22-9-3-1-8(2-4-9)13-7-12(24)15-11(23)5-10(6-14(15)31-13)30-21-18(27)16(25)17(26)19(32-21)20(28)29;/h1-6,13,16-19,21-23,25-27H,7H2,(H,28,29);/q;+1/p-1/t13?,16-,17-,18+,19?,21+;/m0./s1
Formula Weight 470.08
Chemical Name or Material Naringenin 7-O-b-D-Glucuronide Sodium Salt (Mixture of Diastereomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.