missing translation for 'onlineSavingsMsg'
Learn More

Naringenin 4'-O-Beta-D-Glucuronide Sodium Salt (Mixture of Diastereomers), TRC

Product Code. 30467429
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30467429 1 mg 1mg
30466180 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30467429 Supplier Toronto Research Chemicals Supplier No. TRCN3790101MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Naringenin 4'-O-beta-D-Glucuronide Sodium Salt (Mixture of Diastereomers)
Molecular Formula C21H19NaO11
Quantity 1 mg
Synonym 4-[3,4-Dihydro-5,7-dihydroxy-4-oxo-2H-1-benzopyran-2-yl]phenyl β-D-Glucopyranosiduronic Acid Sodium Salt, Naringenin-4'-O-β-D-glucuronopyranoside Sodium Salt
InChI Formula InChI=1S/C21H20O11.Na/c22-9-5-11(23)15-12(24)7-13(31-14(15)6-9)8-1-3-10(4-2-8)30-21-18(27)16(25)17(26)19(32-21)20(28)29;/h1-6,13,16-19,21-23,25-27H,7H2,(H,28,29);/q;+1/p-1/t13?,16-,17-,18+,19-,21+;/m0./s1
SMILES OC1=CC(O)=CC2=C1C(CC(C3=CC=C(O[C@@H]4O[C@H](C(O[Na])=O)[C@@H](O)[C@H](O)[C@H]4O)C=C3)O2)=O
Molecular Weight (g/mol) 470.36
Formula Weight 470.08
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.