missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NARG1L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00
Specifications
| Antigen | NARG1L |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
NARG1L Polyclonal specifically detects NARG1L in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| NARG1L | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 79612 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:FAINSNNPWLHECLIRFSKSVSNHSNLPDIVSKVLSQEMQKIFVKKDLESFNEDFLKRNATSLQHLLSGAKMMYFLDKSRQEKA | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| FLJ22054, MGC40612, N(alpha)-acetyltransferase 16, NatA auxiliary subunit, N-alpha-acetyltransferase 16, NatA auxiliary subunit, NARG1L, NARG1-like protein, NAT2, NMDA receptor regulated 1-like, NMDA receptor-regulated 1-like protein, PRO2435 | |
| NAA16 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title