missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NARG1L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-92166-25ul
This item is not returnable.
View return policy
Description
NARG1L Polyclonal specifically detects NARG1L in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| NARG1L | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| FLJ22054, MGC40612, N(alpha)-acetyltransferase 16, NatA auxiliary subunit, N-alpha-acetyltransferase 16, NatA auxiliary subunit, NARG1L, NARG1-like protein, NAT2, NMDA receptor regulated 1-like, NMDA receptor-regulated 1-like protein, PRO2435 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 79612 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| NAA16 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:FAINSNNPWLHECLIRFSKSVSNHSNLPDIVSKVLSQEMQKIFVKKDLESFNEDFLKRNATSLQHLLSGAKMMYFLDKSRQEKA | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction