missing translation for 'onlineSavingsMsg'
Learn More

Napsin A Mouse anti-Human, Clone: NAPSA/3305, Novus Biologicals™

Mouse Monoclonal Antibody

£377.00

Specifications

Antigen Napsin A
Clone NAPSA/3305
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array
Applications Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18675008
View Documents
Novus Biologicals
NBP3-07866-100ug
100 μg
£377.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Napsin A Monoclonal specifically detects Napsin A in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

Napsin A
0.2 mg/mL
Immunohistochemistry (Paraffin), Peptide Array
Unconjugated
Mouse
Cancer, Cell Biology, Cellular Markers
10 mM PBS with 0.05% BSA
9476
IgG2b κ
Protein A purified
NAPSA/3305
Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array
Monoclonal
Purified
RUO
Human
Asp 4, ASP4, Aspartyl protease 4, EC 3.4.23, EC 3.4.23.-, EC 3.4.23.15, EC 3.4.23.3, EC 3.4.23.5, KAP, Kdap, NAP1napsin-A, NAPApronapsin A, NAPSA, napsin A aspartic peptidase, napsin-1, SNAPA, TA01/TA02
Recombinant human Napsin A protein fragment (aa189-299) (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.