missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Napsin A Mouse anti-Human, Clone: NAPSA/3305, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07866-100ug
This item is not returnable.
View return policy
Description
Napsin A Monoclonal specifically detects Napsin A in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Napsin A | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| Asp 4, ASP4, Aspartyl protease 4, EC 3.4.23, EC 3.4.23.-, EC 3.4.23.15, EC 3.4.23.3, EC 3.4.23.5, KAP, Kdap, NAP1napsin-A, NAPApronapsin A, NAPSA, napsin A aspartic peptidase, napsin-1, SNAPA, TA01/TA02 | |
| Recombinant human Napsin A protein fragment (aa189-299) (exact sequence is proprietary) | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers | |
| 9476 | |
| Store at 4°C. | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Peptide Array | |
| NAPSA/3305 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction