missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPRT1 Mouse anti-Human, Mouse, Clone: 5D8H10, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | NAPRT1 |
|---|---|
| Clone | 5D8H10 |
| Concentration | 1.47 mg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16817785
|
Proteintech
66159-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16807785
|
Proteintech
66159-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nicotinic acid (NA; niacin) is converted by nicotinic acid phosphoribosyltransferase (NAPRT; EC 2.4.2.11) to NA mononucleotide (NaMN), which is then converted to NA adenine dinucleotide (NaAD), and finally to nicotinamide adenine dinucleotide (NAD), which serves as a coenzyme in cellular redox reactions and is an essential component of a variety of processes in cellular metabolism including response to stress (Hara et al., 2007).Specifications
| NAPRT1 | |
| 1.47 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| FHA HIT interacting protein, FHIP, NAPRT1, NAPRTase, PP3856 | |
| NAPRT | |
| IgG2a | |
| Protein A | |
| Antibody |
| 5D8H10 | |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Mouse, Human | |
| Q6XQN6, Q8CC86 | |
| 223646, 93100 | |
| NAPRT1 Fusion Protein Ag4265 | |
| Primary | |
| -20°C | |
| NAPRT |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title