missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPRT1 Mouse anti-Human, Mouse, Clone: 5D8H10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66159-1-IG-150UL
This item is not returnable.
View return policy
Description
Nicotinic acid (NA; niacin) is converted by nicotinic acid phosphoribosyltransferase (NAPRT; EC 2.4.2.11) to NA mononucleotide (NaMN), which is then converted to NA adenine dinucleotide (NaAD), and finally to nicotinamide adenine dinucleotide (NAD), which serves as a coenzyme in cellular redox reactions and is an essential component of a variety of processes in cellular metabolism including response to stress (Hara et al., 2007).Specifications
| NAPRT1 | |
| Monoclonal | |
| 1.47 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6XQN6, Q8CC86 | |
| NAPRT | |
| NAPRT1 Fusion Protein Ag4265 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 5D8H10 | |
| Unconjugated | |
| NAPRT | |
| FHA HIT interacting protein, FHIP, NAPRT1, NAPRTase, PP3856 | |
| Mouse | |
| Protein A | |
| RUO | |
| 223646, 93100 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction