missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£196.00 - £558.00
Specifications
| Antigen | NAPA |
|---|---|
| Dilution | Western Blot 1:500, ELISA 1:100-1:2000 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18027477
|
Novus Biologicals
NBP2-12752-0.025MG |
0.025 mg |
£196.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18095468
|
Novus Biologicals
NBP2-12752-0.05MG |
0.05 mg |
£341.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18192820
|
Novus Biologicals
NBP2-12752 |
0.1 mg |
£558.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NAPA Polyclonal specifically detects NAPA in Human, Mouse, Rat, Porcine, Primate samples. It is validated for Western Blot, ELISA.Specifications
| NAPA | |
| ELISA, Western Blot | |
| Unconjugated | |
| alpha-SNAP, alpha-soluble NSF attachment protein, N-ethylmaleimide-sensitive factor attachment protein alpha, N-ethylmaleimide-sensitive factor attachment protein, alpha, S, SNAPA, SNAP-alpha | |
| NAPA | |
| IgG |
| Western Blot 1:500, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| 8775 | |
| The synthetic peptide taken from human SNAP protein were taken form region corresponding to the aa acid sequences (tmllri kkt iqg dee d). | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title