missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAPA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-12752-0.025MG
This item is not returnable.
View return policy
Description
NAPA Polyclonal specifically detects NAPA in Human, Mouse, Rat, Porcine, Primate samples. It is validated for Western Blot, ELISA.
Specifications
| NAPA | |
| Polyclonal | |
| Unconjugated | |
| Tris/Glycine buffer, pH 7.4-7.8, HEPES,BSA 0.5%, glycerol 30%. with 0.02% Sodium Azide | |
| NAPA | |
| The synthetic peptide taken from human SNAP protein were taken form region corresponding to the aa acid sequences (tmllri kkt iqg dee d). | |
| Affinity Purified | |
| RUO | |
| 8775 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA, Western Blot | |
| 0.50 mg/mL | |
| Western Blot 1:500, ELISA 1:100-1:2000 | |
| alpha-SNAP, alpha-soluble NSF attachment protein, N-ethylmaleimide-sensitive factor attachment protein alpha, N-ethylmaleimide-sensitive factor attachment protein, alpha, S, SNAPA, SNAP-alpha | |
| Rabbit | |
| 36 kDa | |
| 0.025 mg | |
| Primary | |
| Pig, Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction